accessibility menu, dialog, popup

UserName

CAS RN 120-32-1

OHCl

CAS RN 120-32-1

IUPAC Name: 2-benzyl-4-chlorophenol
Synonyms: chlorophene caswell no. 083 septiphene neosabenyl orthosan santophen 1 solution clorofenum o-cresol, 4-chloro-α-phenyl- epa pesticide chemical code 062201 clorophene potassium salt
Molecular Weight (g/mol): 218.68
Molecular Formula: C13H11ClO
InChi Key: NCKMMSIFQUPKCK-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)