accessibility menu, dialog, popup

UserName

CAS RN 1198-51-2

IUPAC Name: 3-(bromomethyl)-5-chloro-1-benzothiophene
Synonyms: 3-(bromomethyl)-5-chloro-1-benzothiophene
Molecular Weight (g/mol): 261.56
Molecular Formula: C9H6BrClS
InChi Key: FKQSFVITUNJLCY-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SC=C(CBr)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)