accessibility menu, dialog, popup

UserName

H2OH2OH2OH3C(S)OH(R)CH3NOHOOS(S)HN(S)ONCH3CH3

CAS RN 119478-56-7

IUPAC Name: (4R)-3-{[(3S,5S)-5-(dimethylcarbamoyl)pyrrolidin-3-yl]sulfanyl}-6-[(1S)-1-hydroxyethyl]-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid trihydrate
Synonyms: meropenem
Molecular Weight (g/mol): 437.51
Molecular Formula: C17H31N3O8S
InChi Key: CTUAQTBUVLKNDJ-TXBRDXQXSA-N
SMILES: O.O.O.C[C@H](O)C1C2[C@@H](C)C(S[C@@H]3CN[C@@H](C3)C(=O)N(C)C)=C(N2C1=O)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (7)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (1)
Color 
  • (2)
  • (1)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (7)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)