accessibility menu, dialog, popup

UserName

CAS RN 1193-92-6

OHNN(E)

CAS RN 1193-92-6

IUPAC Name: (E)-N-[(pyridin-3-yl)methylidene]hydroxylamine
Synonyms: (E)-N-(pyridin-3-ylmethylidene)hydroxylamine
Molecular Weight (g/mol): 122.13
Molecular Formula: C6H6N2O
InChi Key: YBKOPFQCLSPTPV-VMPITWQZSA-N
SMILES: O\N=C\C1=CC=CN=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)