accessibility menu, dialog, popup

UserName

H3COH

CAS RN 1193-81-3

IUPAC Name: 1-cyclohexylethan-1-ol
Synonyms: 1-cyclohexylethanol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: JMSUNAQVHOHLMX-UHFFFAOYNA-N
SMILES: CC(O)C1CCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results