accessibility menu, dialog, popup

UserName

CAS RN 1193-21-1

ClClNN

CAS RN 1193-21-1

IUPAC Name: 4,6-dichloropyrimidine
Synonyms: 4,6-dichloropyrimidine
Molecular Weight (g/mol): 148.97
Molecular Formula: C4H2Cl2N2
InChi Key: XJPZKYIHCLDXST-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=NC=N1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)