accessibility menu, dialog, popup

UserName

CAS RN 1192758-40-9

ONNNOOHH2OHCl

CAS RN 1192758-40-9

IUPAC Name: 6-(morpholin-4-yl)pyridazine-3-carboxylic acid hydrate hydrochloride
Synonyms: 6-(morpholin-4-yl)pyridazine-3-carboxylic acid hydrate hydrochloride
Molecular Weight (g/mol): 263.68
Molecular Formula: C9H14ClN3O4
InChi Key: VARLCSQEQQKXKH-UHFFFAOYSA-N
SMILES: O.Cl.OC(=O)C1=NN=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)