accessibility menu, dialog, popup

UserName

FNSH2NF

CAS RN 119256-40-5

IUPAC Name: 4,6-difluoro-1,3-benzothiazol-2-amine
Synonyms: 4,6-difluoro-1,3-benzothiazol-2-amine
Molecular Weight (g/mol): 186.18
Molecular Formula: C7H4F2N2S
InChi Key: DDKKXSCVPKDRRS-UHFFFAOYSA-N
SMILES: NC1=NC2=C(F)C=C(F)C=C2S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results