accessibility menu, dialog, popup

UserName

CAS RN 1192-30-9

BrO

CAS RN 1192-30-9

IUPAC Name: 2-(bromomethyl)oxolane
Synonyms: 2-(bromomethyl)oxolane
Molecular Weight (g/mol): 165.03
Molecular Formula: C5H9BrO
InChi Key: VOHILFSOWRNVJJ-UHFFFAOYNA-N
SMILES: BrCC1CCCO1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)