accessibility menu, dialog, popup

UserName

CAS RN 1191-99-7

O

CAS RN 1191-99-7

IUPAC Name: 2,3-dihydrofuran
Synonyms: 2,3-dihydrofuran
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: JKTCBAGSMQIFNL-UHFFFAOYSA-N
SMILES: C1CC=CO1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (5)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)