accessibility menu, dialog, popup

UserName

CH3OOH3CH3C

CAS RN 1191-16-8

IUPAC Name: 3-methylbut-2-en-1-yl acetate
Synonyms: prenyl acetate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: XXIKYCPRDXIMQM-UHFFFAOYSA-N
SMILES: CC(C)=CCOC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results