accessibility menu, dialog, popup

UserName

CH3CH3OHCH3CH3H3COHH3CH3CCH3

CAS RN 119-47-1

IUPAC Name: 2-tert-butyl-6-[(3-tert-butyl-2-hydroxy-5-methylphenyl)methyl]-4-methylphenol
Synonyms: cyanox 2246 methylene di-T-butylcresol
Molecular Weight (g/mol): 340.51
Molecular Formula: C23H32O2
InChi Key: KGRVJHAUYBGFFP-UHFFFAOYSA-N
SMILES: CC1=CC(CC2=CC(C)=CC(=C2O)C(C)(C)C)=C(O)C(=C1)C(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results