accessibility menu, dialog, popup

UserName

CAS RN 119-32-4

IUPAC Name: 4-methyl-3-nitroaniline
Synonyms: 3-nitro-4-toluidine
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: GDIIPKWHAQGCJF-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C=C1[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results