CAS RN 1189865-88-0
CAS RN 1189865-88-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
rac Didemethyl Citalopram-d6 Hydrochloride, TRC
CAS: 1189865-88-0 Molecular Formula: C18H12D6ClFN2O Molecular Weight (g/mol): 338.84 SMILES: Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C1(OCc2cc(ccc12)C#N)c3ccc(F)cc3