CAS RN 118876-58-7
CAS RN 118876-58-7
IUPAC Name:
6-nitro-2,3-dioxo-1H,2H,3H,4H-benzo[f]quinoxaline-7-sulfonamide
Synonyms:
NBQX
Molecular Weight (g/mol):
336.28
Molecular Formula:
C12H8N4O6S
InChi Key:
UQNAFPHGVPVTAL-UHFFFAOYSA-N
SMILES:
NS(=O)(=O)C1=C2C(=CC=C1)C1=C(NC(=O)C(=O)N1)C=C2[N+]([O-])=O
1
–
4
of
4
results
2
Tocris Bioscience™ NBQX
CAS: 118876-58-7 Molecular Formula: C12H8N4O6S
3
6-Nitro-7-sulfamoylbenzo[f]quinoxaline-2,3-Dione, TRC
CAS: 118876-58-7 Molecular Formula: C12 H8 N4 O6 S Molecular Weight (g/mol): 336.28 Synonym: 6-Nitro-7-sulfamoylbenzo[f]quinoxaline-2,3-dione,NBQX IUPAC Name: 6-nitro-2,3-dioxo-1,4-dihydrobenzo[f]quinoxaline-7-sulfonamide SMILES: NS(=O)(=O)c1cccc2c3NC(=O)C(=O)Nc3cc(c12)[N+](=O)[O-]
4
NBQX, MedChemExpress
MedChemExpress NBQX (FG9202) is a highly selective and competitive AMPA receptor antagonist. NBQX has neuroprotective and anticonvulsant activity.