accessibility menu, dialog, popup

UserName

CAS RN 118803-40-0

IUPAC Name: 6-nitro-1,3-dihydro-2,1-benzoxaborol-1-ol
Synonyms: 6-nitro-3H-2,1-benzoxaborol-1-ol
Molecular Weight (g/mol): 178.94
Molecular Formula: C7H6BNO4
InChi Key: GFFKBQCIGADRSN-UHFFFAOYSA-N
SMILES: OB1OCC2=C1C=C(C=C2)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results