accessibility menu, dialog, popup

UserName

CAS RN 1187236-18-5

H3COONNBr

CAS RN 1187236-18-5

IUPAC Name: ethyl 7-bromoimidazo[1,2-a]pyridine-2-carboxylate
Synonyms: ethyl 7-bromoimidazo[1,2-a]pyridine-2-carboxylate
Molecular Weight (g/mol): 269.1
Molecular Formula: C10H9BrN2O2
InChi Key: ZNBQFCWFNJYWEE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN2C=CC(Br)=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results