accessibility menu, dialog, popup

UserName

NH2NH2NN(Z)

CAS RN 1187-42-4

IUPAC Name: (2Z)-diaminobut-2-enedinitrile
Synonyms: (2Z)-diaminobut-2-enedinitrile
Molecular Weight (g/mol): 108.1
Molecular Formula: C4H4N4
InChi Key: DPZSNGJNFHWQDC-ARJAWSKDSA-N
SMILES: N\C(C#N)=C(/N)C#N
Molecular Weight (g/mol) 
  • (6)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)