accessibility menu, dialog, popup

UserName

CAS RN 1186405-11-7

NNFBr

CAS RN 1186405-11-7

IUPAC Name: 3-bromo-6-fluoroimidazo[1,2-a]pyridine
Synonyms: 3-bromo-6-fluoroimidazo[1,2-a]pyridine
Molecular Weight (g/mol): 215.03
Molecular Formula: C7H4BrFN2
InChi Key: HGFXJQASNPSBAZ-UHFFFAOYSA-N
SMILES: FC1=CN2C(Br)=CN=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results