CAS RN 1186-09-0
CAS RN 1186-09-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
O,O,S-Triethyl Phosphorothiolate, TRC
CAS: 1186-09-0 Molecular Formula: C6 H15 O3 P S Molecular Weight (g/mol): 198.22 Synonym: Phosphorothioic acid, O,O,S-triethyl ester,Ethyl phosphorothioate ((EtO)2(EtS)PO) (6CI),Ethyl thiophosphate ((EtO)2(EtS)PO) (5CI),O,O,S-Triethyl phosphorothioate,O,O,S-Triethyl phosphorothiolate,O,O,S-Triethyl thiophosphate,O,O-Diethyl S-ethyl phosphorothioate,Thiophosphoric acid O,O,S-triethyl ester,Triethyl phosphorothioate IUPAC Name: 1-[ethoxy(ethylsulfanyl)phosphoryl]oxyethane SMILES: CCOP(=O)(OCC)SCC