accessibility menu, dialog, popup

UserName

CAS RN 118430-73-2

CH3NNH2NCH3H3CCH3

CAS RN 118430-73-2

IUPAC Name: 3-tert-butyl-1-methyl-1H-pyrazol-5-amine
Synonyms: 5-tert-butyl-2-methylpyrazol-3-amine
Molecular Weight (g/mol): 153.23
Molecular Formula: C8H15N3
InChi Key: XSCDSAMVQLKDNI-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1N)C(C)(C)C
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)