accessibility menu, dialog, popup

UserName

CAS RN 118-13-8

HOOHON

CAS RN 118-13-8

IUPAC Name: 3-hydroxyquinoline-4-carboxylic acid
Synonyms: cinchoninic acid, 3-hydroxy-
Molecular Weight (g/mol): 189.17
Molecular Formula: C10H7NO3
InChi Key: MXNVEJDRXSFZQB-UHFFFAOYSA-N
SMILES: OC(=O)C1=C2C=CC=CC2=NC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results