accessibility menu, dialog, popup

UserName

CH3OCH3

CAS RN 116903-46-9

IUPAC Name: 1-ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene
Synonyms: 1-ethoxy-4-[2-(4-methylphenyl)ethynyl]benzene
Molecular Weight (g/mol): 236.31
Molecular Formula: C17H16O
InChi Key: DURDZFYGFXPFAV-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C#CC1=CC=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results