accessibility menu, dialog, popup

UserName

CAS RN 116855-09-5

IUPAC Name: 1-methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid
Synonyms: 1-methylpyrazolo[3,4-b]pyridine-3-carboxylic acid
Molecular Weight (g/mol): 177.16
Molecular Formula: C8H7N3O2
InChi Key: WEZYUZRHWLFJDN-UHFFFAOYSA-N
SMILES: CN1N=C(C(O)=O)C2=CC=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results