accessibility menu, dialog, popup

UserName

CAS RN 116750-06-2

ONOOO

CAS RN 116750-06-2

IUPAC Name: 5-[(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)methyl]furan-2-carbaldehyde
Synonyms: 5-[(1,3-dioxoisoindol-2-yl)methyl]furan-2-carbaldehyde
Molecular Weight (g/mol): 255.23
Molecular Formula: C14H9NO4
InChi Key: ZRBXPAHJXKKXME-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2C(=O)C3=CC=CC=C3C2=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results