accessibility menu, dialog, popup

UserName

HNNH2HClHCl

CAS RN 116183-83-6

IUPAC Name: pyrrolidin-3-amine dihydrochloride
Synonyms: pyrrolidin-3-amine dihydrochloride
Molecular Weight (g/mol): 159.05
Molecular Formula: C4H12Cl2N2
InChi Key: NJPNCMOUEXEGBL-UHFFFAOYNA-N
SMILES: Cl.Cl.NC1CCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results