accessibility menu, dialog, popup

UserName

NH2OHOO

CAS RN 116-85-8

IUPAC Name: 1-amino-4-hydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: 1-amino-4-hydroxyanthraquinone disperse red 15, except as impurity in disperse violet 1
Molecular Weight (g/mol): 239.23
Molecular Formula: C14H9NO3
InChi Key: AQXYVFBSOOBBQV-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)