accessibility menu, dialog, popup

UserName

CAS RN 116-63-2

NH2HOSOHOO

CAS RN 116-63-2

IUPAC Name: 4-amino-3-hydroxynaphthalene-1-sulfonic acid
Synonyms: 1-amino-4-sulfo-2-naphthol
Molecular Weight (g/mol): 239.25
Molecular Formula: C10H9NO4S
InChi Key: RXCMFQDTWCCLBL-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
Molecular Weight (g/mol) 
  • (5)
  • (7)
Quantity 
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (5)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (5)
  • (3)
  • (1)
  • (5)
  • (7)
  • (4)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)