accessibility menu, dialog, popup

UserName

CAS RN 116-53-0

CH3CH3OHO

CAS RN 116-53-0

IUPAC Name: 2-methylbutanoic acid
Synonyms: (+-)-2-methylbutyrate active valeric acid DL-2-methy butyric acid sodium salt 2-methylbutyrate DL-2-methy butyrate 2-methylbutyrate, (+-)-isomer rs-2-methylbutyrate (+-)-isomer 2-methylbutyrate 2-methylbutyric acid (natrual) valeric acid, active
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: WLAMNBDJUVNPJU-UHFFFAOYNA-N
SMILES: CCC(C)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results