accessibility menu, dialog, popup

UserName

HOOOHNSOONHSN

CAS RN 116-43-8

IUPAC Name: 3-({4-[(1,3-thiazol-2-yl)sulfamoyl]phenyl}carbamoyl)propanoic acid
Synonyms: succinylsulfathiazole kaoxidine sulfadigesin thiacyl sulfasuccinil sulfasuccithiazole succinylsulfathiazol sulfasuxidine 琥珀磺胺噻唑 sulfasuccidin
Molecular Weight (g/mol): 355.38
Molecular Formula: C13H13N3O5S2
InChi Key: SKVLYVHULOWXTD-UHFFFAOYSA-N
SMILES: OC(=O)CCC(=O)NC1=CC=C(C=C1)S(=O)(=O)NC1=NC=CS1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)