CAS RN 115960-14-0
CAS RN 115960-14-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hidiosmin, TRC
CAS: 115960-14-0 Molecular Formula: C30H36O16 Molecular Weight (g/mol): 652.6 Synonym: 7-[[6-O-(6-Deoxy-alpha-L-mannopyranosyl)-beta-D-glucopyranosyl]oxy]-5-(2-hydroxyethoxy)-2-(3-hydroxy-4-methoxyphenyl)-4H-1-benzopyran-4-one,5-O-(beta-Hydroxyethyl)diosmin IUPAC Name: 5-(2-hydroxyethoxy)-2-(3-hydroxy-4-methoxyphenyl)-7-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one SMILES: COc1ccc(cc1O)C2=CC(=O)c3c(OCCO)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2