accessibility menu, dialog, popup

UserName

CAS RN 115833-93-7

IUPAC Name: 3-(pyrrolidin-1-yl)aniline
Synonyms: 3-(pyrrolidin-1-yl)aniline
Molecular Weight (g/mol): 162.24
Molecular Formula: C10H14N2
InChi Key: DNPBFTQZONVQDX-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results