accessibility menu, dialog, popup

UserName

CAS RN 115473-15-9

NHOSHCl

CAS RN 115473-15-9

IUPAC Name: 2H,4H,5H,6H,7H,7aH-thieno[3,2-c]pyridin-2-one hydrochloride
Synonyms: 4H,5H,6H,7H,7aH-thieno[3,2-c]pyridin-2-one hydrochloride
Molecular Weight (g/mol): 191.67
Molecular Formula: C7H10ClNOS
InChi Key: PUQKTVAKLPDUAW-UHFFFAOYNA-N
SMILES: Cl.O=C1SC2CCNCC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results