accessibility menu, dialog, popup

UserName

CAS RN 115377-94-1

IUPAC Name: (2-{[(tert-butoxy)carbonyl]amino}phenyl)boronic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]phenylboronic acid
Molecular Weight (g/mol): 237.06
Molecular Formula: C11H16BNO4
InChi Key: RMEIKYSECODAHJ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NC1=CC=CC=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results