accessibility menu, dialog, popup

UserName

CAS RN 1151512-19-4

SOCl

CAS RN 1151512-19-4

IUPAC Name: 4-[(thiophen-2-yl)methyl]benzoyl chloride
Synonyms: 4-(thiophen-2-ylmethyl)benzoyl chloride
Molecular Weight (g/mol): 236.71
Molecular Formula: C12H9ClOS
InChi Key: AOHWXUBTRWZCOZ-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=C(CC2=CC=CS2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results