CAS RN 1150-90-9
CAS RN 1150-90-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Estratetraenol, TRC
CAS: 1150-90-9 Molecular Formula: C18 H22 O Molecular Weight (g/mol): 254.37 Synonym: Estra-1,3,5(10),16-tetraen-3-ol,Compound 742 IUPAC Name: (8S,9S,13R,14S)-13-methyl-6,7,8,9,11,12,14,15-octahydrocyclopenta[a]phenanthren-3-ol SMILES: C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC=C2