accessibility menu, dialog, popup

UserName

CH3CH3H3COH3COCH2

CAS RN 115-95-7

IUPAC Name: 3,7-dimethylocta-1,6-dien-3-yl acetate
Synonyms: linalyl acetate
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: UWKAYLJWKGQEPM-UHFFFAOYNA-N
SMILES: CC(C)=CCCC(C)(OC(C)=O)C=C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)