accessibility menu, dialog, popup

UserName

HClHONH

CAS RN 115-46-8

IUPAC Name: hydrogen diphenyl(piperidin-4-yl)methanol chloride
Synonyms: azacyclonol hydrochloride azacyclonol (inn) and its salts
Molecular Weight (g/mol): 303.83
Molecular Formula: C18H22ClNO
InChi Key: LBRAWKGGVUCSOX-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(C1CCNCC1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (9)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results