accessibility menu, dialog, popup

UserName

H3CBrOHOSOOCH3HOBr

CAS RN 115-40-2

IUPAC Name: 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: bromocresol purple bromcresol purple
Molecular Weight (g/mol): 540.22
Molecular Formula: C21H16Br2O5S
InChi Key: ABIUHPWEYMSGSR-UHFFFAOYSA-N
SMILES: CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (6)
  • (1)
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (18)
Grade 
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
Color 
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
pH 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (6)
  • (1)
  • (5)
  • (1)
  • (1)
  • (18)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (3)