accessibility menu, dialog, popup

UserName

OHBrBrOSOOBrHOBr

CAS RN 115-39-9

IUPAC Name: 3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Synonyms: bromophenol blue bromophenol blue, sultone form
Molecular Weight (g/mol): 669.96
Molecular Formula: C19H10Br4O5S
InChi Key: UDSAIICHUKSCKT-UHFFFAOYSA-N
SMILES: OC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
Quantity 
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (10)
  • (1)
  • (8)
  • (3)
  • (1)
Grade 
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
Color 
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
pH 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (7)
  • (5)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (10)
  • (1)
  • (8)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)