accessibility menu, dialog, popup

UserName

HOClClCl

CAS RN 115-20-8

IUPAC Name: 2,2,2-trichloroethan-1-ol
Synonyms: trichloroethanol trichloroethyl alcohol (substance) C2H3cl3O
Molecular Weight (g/mol): 149.4
Molecular Formula: C2H3Cl3O
InChi Key: KPWDGTGXUYRARH-UHFFFAOYSA-N
SMILES: OCC(Cl)(Cl)Cl
Boiling Point 
  • (4)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)