accessibility menu, dialog, popup

UserName

2H2H2H2H2H2H2H2H

CAS RN 1146-65-2

IUPAC Name: (²H₈)naphthalene
Synonyms: (²H₈)naphthalene
Molecular Weight (g/mol): 136.22
Molecular Formula: C10H8
InChi Key: UFWIBTONFRDIAS-PGRXLJNUSA-N
SMILES: [2H]C1=C([2H])C2=C([2H])C([2H])=C([2H])C([2H])=C2C([2H])=C1[2H]
Quantity 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results