CAS RN 1145-93-3
IUPAC Name:
ethoxy ethyl [(4-methoxyphenyl)methyl]phosphonite
Synonyms:
ethoxy ethyl (4-methoxyphenyl)methylphosphonite
Molecular Weight (g/mol):
258.25
Molecular Formula:
C12H19O4P
InChi Key:
XTASVOIVNCSQLW-UHFFFAOYNA-N
SMILES:
CCOOP(CC1=CC=C(OC)C=C1)OCC
1
–
1
of
1
results
1
Diethyl 4-methoxybenzylphosphonate, 98+%
CAS: 1145-93-3 Molecular Formula: C12H19O4P Molecular Weight (g/mol): 258.254 MDL Number: MFCD00015662 InChI Key: NKARVHNAACNYGE-UHFFFAOYSA-N Synonym: diethyl 4-methoxybenzylphosphonate,diethyl 4-methoxybenzyl phosphonate,1-diethoxyphosphorylmethyl-4-methoxybenzene,diethyl 4-methoxyphenyl methylphosphonate,1-diethoxyphosphorylmethyl-4-methoxy-benzene,phosphonic acid, 4-methoxyphenyl methyl-, diethyl ester,acmc-1bsuk,4-diethylphosphonomethyl anisole,diethyl-4-methoxybenzyl phosphonate,diethyl 4-methoxybenzyl-phosphonate PubChem CID: 4063826 IUPAC Name: 1-(diethoxyphosphorylmethyl)-4-methoxybenzene SMILES: CCOP(=O)(CC1=CC=C(C=C1)OC)OCC