accessibility menu, dialog, popup

UserName

CAS RN 1142-15-0

IUPAC Name: 1-methoxy-4-[(1E)-2-phenylethenyl]benzene
Synonyms: p-methoxystilbene
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: XWYXLYCDZKRCAD-BQYQJAHWSA-N
SMILES: COC1=CC=C(\C=C\C2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results