CAS RN 113893-08-6
CAS RN 113893-08-6
IUPAC Name:
(1-benzothiophen-3-yl)boronic acid
Synonyms:
1-benzothiophen-3-ylboronic acid
Molecular Weight (g/mol):
178.01
Molecular Formula:
C8H7BO2S
InChi Key:
QVANIYYVZZLQJP-UHFFFAOYSA-N
SMILES:
OB(O)C1=CSC2=CC=CC=C12
1
–
3
of
3
results
1
Benzo[b]thien-3-ylboronic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Benzo[b]thien-3-ylboronic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Benzo[b]thien-3-ylboronic Acid, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.