missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thien-3-ylboronic Acid, TRC

Product Code. 30344729
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344729 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344729 Supplier Toronto Research Chemicals Supplier No. TRCB2068905G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzo[b]thien-3-ylboronic Acid
CAS 113893-08-6
Molecular Formula C8H7BO2S
Quantity 5 g
Synonym (1-Benzothien-3-yl)boronic Acid, (Benzo[b]thien-3-yl)boronic Acid, (Benzo[b]thiophen-3-yl)boronic Acid, (Benzothiophen-3-yl)boronic Acid, (Benzothiophene-3-yl)boronic Acid, 3-Benzo[b]thienylboronic Acid, Benzothien-3-ylboronic Acid, Benzothiophene-3-boronic Acid, Thianaphthen-3-ylboronic Acid
InChI Formula InChI=1S/C8H7BO2S/c10-9(11)7-5-12-8-4-2-1-3-6(7)8/h1-5,10-11H
SMILES OB(O)c1csc2ccccc12
IUPAC Name 1-benzothiophen-3-ylboronic acid
Molecular Weight (g/mol) 178.02
Formula Weight 178.026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.