accessibility menu, dialog, popup

UserName

CAS RN 1138-15-4

ONN(E)

CAS RN 1138-15-4

IUPAC Name: (2E)-1-(1H-imidazol-1-yl)-3-phenylprop-2-en-1-one
Synonyms: (2E)-1-(imidazol-1-yl)-3-phenylprop-2-en-1-one
Molecular Weight (g/mol): 198.23
Molecular Formula: C12H10N2O
InChi Key: XVGXMXZUJNAGFZ-VOTSOKGWSA-N
SMILES: O=C(\C=C\C1=CC=CC=C1)N1C=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results