accessibility menu, dialog, popup

UserName

CAS RN 113732-84-6

CH3NOO

CAS RN 113732-84-6

IUPAC Name: 2-methyl-1,3-oxazole-4-carbaldehyde
Synonyms: 2-methyl-1,3-oxazole-4-carbaldehyde
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: ARAUEWKXKTYCHZ-UHFFFAOYSA-N
SMILES: CC1=NC(C=O)=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results