accessibility menu, dialog, popup

UserName

CH2NONHSS

CAS RN 113520-01-7

IUPAC Name: 11-(prop-2-en-1-yl)-10-sulfanylidene-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6)-dien-12-one
Synonyms: 11-(prop-2-en-1-yl)-10-sulfanylidene-7-thia-9,11-diazatricyclo[6.4.0.0²,⁶]dodeca-1(8),2(6)-dien-12-one
Molecular Weight (g/mol): 264.36
Molecular Formula: C12H12N2OS2
InChi Key: SRKGQOMPUYHFAP-UHFFFAOYSA-N
SMILES: C=CCN1C(=S)NC2=C(C3=C(CCC3)S2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results