accessibility menu, dialog, popup

UserName

CAS RN 113512-71-3

OHFFNH2

CAS RN 113512-71-3

IUPAC Name: 5-amino-2,4-difluorophenol
Synonyms: 5-amino-2,4-difluorophenol
Molecular Weight (g/mol): 145.11
Molecular Formula: C6H5F2NO
InChi Key: CXADAFQCFSVRGN-UHFFFAOYSA-N
SMILES: NC1=CC(O)=C(F)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results